Learn More
Medchemexpress LLC Ribociclib-d6 hydrochloride | 1211443-80-9 | 98.3% | 477.04 g/mol | C23H25D6ClN8O | 10 MG
SDP
Catalog No. 5000288423
Encompass_Preferred
Ribociclib-d6 hydrochloride is the deuterium-labeled hydrochloride salt of ribociclib, provided as a solid reference standard for analytical, pharmacokinetic, and bioanalytical research requiring isotopically labeled material.
  • Deuterium-labeled internal standard for quantitative mass spectrometry
  • High purity (~98.3%) suitable for analytical reference use
  • Solid form, light yellow to light brown in appearance
  • Storage recommendations: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month)
  • Appropriate for pharmacokinetic, metabolic, and method development studies
Catalog No. 50-002-88423 Supplier Medchemexpress LLC Supplier No. HY15777AS10MG
Ribociclib-d6 hydrochloride is the deuterium-labeled hydrochloride salt of ribociclib, provided as a solid reference standard for analytical, pharmacokinetic, and bioanalytical research requiring isotopically labeled material.
  • Deuterium-labeled internal standard for quantitative mass spectrometry
  • High purity (~98.3%) suitable for analytical reference use
  • Solid form, light yellow to light brown in appearance
  • Storage recommendations: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month)
  • Appropriate for pharmacokinetic, metabolic, and method development studies