Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribociclib-d8 | 2167898-24-8 | 99.2% | 440.59 | C23H25D8N8O | 10 MG
SDP

Supplier:  Medchemexpress LLC HY15777S110MG

Encompass_Preferred

Ribociclib-d8 is the deuterium-labeled analogue of ribociclib supplied as an analytical reference standard for use in mass spectrometry and quantitative assays. It is intended for research use and provides a traceable internal standard for accurate quantitation in bioanalytical workflows.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity suitable for analytical applications.
  • Soluble in DMSO for convenient preparation of stock solutions.
  • Molecular weight 440.59 and formula C23H25D8N8O for exact mass matching.
  • Provided in a 10 MG quantity appropriate for method development and validation.

Catalog No. 50-002-88335


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.