Learn More
Selleck Chemical LLC Ribociclib hydrochloride S5187-1g
SDP
Catalog No. 503111990
Encompass_Preferred
Ribociclib (LEE011) hydrochloride is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively
Catalog No. 50-311-1990 Supplier Selleck Chemical LLC Supplier No. S51871G
Ribociclib (LEE011) hydrochloride is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively