Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribociclib succinate | 1374639-75-4 | MFCD28167746 | 99.9% | 552.63 g/mol | C27H36N8O5 | 10 MG
SDP

Catalog No. 5000287928
Encompass_Preferred

Ribociclib succinate is the succinate salt form of ribociclib (LEE011), a selective cyclin-dependent kinase 4/6 (CDK4/6) inhibitor for research and analytical use. It is used in preclinical biochemical and cell-based studies to investigate cell-cycle regulation and oncology-related pathways.

  • Selective CDK4/6 inhibitor with reported IC50 values of 10 nM and 39 nM.
  • High purity suitable for research; example certificate of analysis lists 99.93% purity.
  • Molecular formula C27H36N8O5 and molecular weight 552.63 g/mol.
  • Supplied in milligram quantities to support laboratory experiments.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-87928 Supplier Medchemexpress LLC Supplier No. HY15777B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ribociclib succinate is the succinate salt form of ribociclib (LEE011), a selective cyclin-dependent kinase 4/6 (CDK4/6) inhibitor for research and analytical use. It is used in preclinical biochemical and cell-based studies to investigate cell-cycle regulation and oncology-related pathways.

  • Selective CDK4/6 inhibitor with reported IC50 values of 10 nM and 39 nM.
  • High purity suitable for research; example certificate of analysis lists 99.93% purity.
  • Molecular formula C27H36N8O5 and molecular weight 552.63 g/mol.
  • Supplied in milligram quantities to support laboratory experiments.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.