Learn More
Selleck Chemical LLC Ribociclib succinate S5188-10mM/1mL
SDP
Catalog No. 503112201
Encompass_Preferred
Ribociclib (LEE011) succinate is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively
Catalog No. 50-311-2201 Supplier Selleck Chemical LLC Supplier No. S518810MM/1ML
Ribociclib (LEE011) succinate is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively