Learn More
Selleck Chemical LLC Ribociclib succinate S5188-25mg
SDP
Catalog No. 503112802
Encompass_Preferred
Ribociclib (LEE011) succinate is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively
Catalog No. 50-311-2802 Supplier Selleck Chemical LLC Supplier No. S518825MG
Ribociclib (LEE011) succinate is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively