Learn More
Selleck Chemical LLC Ribociclib succinate S5188-5mg
SDP
Catalog No. 503112258
Encompass_Preferred
Ribociclib (LEE011) succinate is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively
Catalog No. 50-311-2258 Supplier Selleck Chemical LLC Supplier No. S51885MG
Ribociclib (LEE011) succinate is a highly specific dual inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM respectively