Learn More
Medchemexpress LLC HY-19488B 1mg Medchemexpress, Ribocil-C R enantiomer CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19488B1MG
Encompass_Preferred
Medchemexpress, HY-19488B 1mg Ribocil-C R enantiomer CAS: Ribocil-C R enantiomer is the R enantiomer of Ribocil-C. Ribocil-C is a highly selective inhibitor of bacterial riboflavin riboswitches. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2834