Learn More
Sigma Aldrich Fine Chemicals Biosciences Riboflavin 1KG
Catalog No. 502907715
Encompass
Riboflavin is an essential vitamin soluble in water. Milk and eggs are found to be its major sources. Riboflavin is usually bound to enzymes in all tissues. During riboflavin localization in plasma it is bound to albumin and specific immunoglobulins.
Catalog No. 50-290-7715 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R76491KG
Add to Cart
Add to Cart
Riboflavin is an essential vitamin soluble in water. Milk and eggs are found to be its major sources. Riboflavin is usually bound to enzymes in all tissues. During riboflavin localization in plasma it is bound to albumin and specific immunoglobulins.