Learn More
Sigma Aldrich Fine Chemicals Biosciences Riboflavin 5 monophosphate10MG
Catalog No. 502904140
Encompass
Riboflavin 5AE-monophosphate (fMN) is highly soluble in water. flavins are considered as blue-light absorbing dyes.
Catalog No. 50-290-4140 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F839910MG
Riboflavin 5AE-monophosphate (fMN) is highly soluble in water. flavins are considered as blue-light absorbing dyes.