Learn More
Cayman Chemical Riboflavin 5'-monophosphate (sodium salt hydrate), Purity: ≥95%, CAS: 6184-17-4, 100 g
SDP
Catalog No. 502966674
Encompass_Preferred
A coenzyme that is tightly bound to enzymes catalyzing oxidation and reduction reactions in a variety of biosynthetic pathways; binds the FMN riboswitch (RFN element) on RNA to alter gene regulation; a substrate of FMN phosphohydrolases
Catalog No. 50-296-6674 Supplier Cayman Chemical Supplier No. 18167100 G
A coenzyme that is tightly bound to enzymes catalyzing oxidation and reduction reactions in a variety of biosynthetic pathways; binds the FMN riboswitch (RFN element) on RNA to alter gene regulation; a substrate of FMN phosphohydrolases