Learn More
Abcam Riboflavin-5-phosphate sodium salt dihydrate, Enzyme cofactor, 25G
Catalog No. 502704694
Encompass_Preferred
MW 515.4 g/mol. Enzyme cofactor used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Also used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. Can be used as a photoinitiator of polymerization for PAGE gels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4694 Supplier Abcam Supplier No. AB14634925G
MW 515.4 g/mol. Enzyme cofactor used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Also used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. Can be used as a photoinitiator of polymerization for PAGE gels.
The product is subject to the following: Abcam Restricted Use Statement