Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Riboflavin-5'-phosphate sodium salt dihydrate

Click to view available options
Quantity:
10g
50g
6184-17-4
C17H25N4NaO11P
515.368
MFCD00150993
KQNFOSVYQPLXAT-QPVGXXQTSA-N
riboflavin 5'-monophosphate sodium salt dihydrate
131856174
[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
CC1=CC2=C(C=C1C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(COP(=O)(O)O)O)O)O.O.O.[Na]
6184-17-4
C17H25N4NaO11P
515.368
MFCD00150993
KQNFOSVYQPLXAT-QPVGXXQTSA-N
riboflavin 5'-monophosphate sodium salt dihydrate
131856174
[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
CC1=CC2=C(C=C1C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(COP(=O)(O)O)O)O)O.O.O.[Na]
Quantity 50g
Melting Point >300°C
Color Yellow to Orange
Beilstein 4106529
Merck Index 14,8201
Solubility Information Soluble in water at 50mg/mL
Formula Weight 514.36 (478.34 Anhydrous)
Physical Form Powder
Chemical Name or Material Riboflavin-5'-phosphate sodium salt dihydrate

TSCA : TSCA

storageNote1 : 2° to 8°C