Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Riboflavin-5-phosphate, Sodium salt, MP Biomedicals™

Catalog No. ICN10281605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10281605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10281605 Supplier MP Biomedicals, Inc Supplier No. 0210281605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

USP grade photoinitiator for polymerization in acid or alkaline buffer systems.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-40-5
Molecular Formula C17H21N4NaO9P
Molecular Weight (g/mol) 479.34
MDL Number MFCD00065362,MFCD00150992
InChI Key WXEASYCOEACRIK-UHFFFAOYNA-N
PubChem CID 131673928
IUPAC Name [(5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl)oxy]phosphonic acid sodium
SMILES [Na].CC1=C(C)C=C2N(CC(O)C(O)C(O)COP(O)(O)=O)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Color Orange
Quantity 5 g
Formula Weight 478.3
Grade Not Available
Physical Form Powder
Chemical Name or Material Riboflavin-5-phosphate, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.