Learn More
Medchemexpress LLC Riboflavin-d3 | 83-88-5 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0456S31MG
Encompass_Preferred
Riboflavin-d3 is a deuterium-labeled form of Riboflavin (Vitamin B2). Riboflavin is an easily absorbed micronutrient essential for maintaining health in humans and other animals. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis. Deuterium substitution has been noted for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06428