Learn More
Medchemexpress LLC Riboflavin-d3 (Vitamin B2-d3) | 83-88-5 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0456S35MG
Encompass_Preferred
Riboflavin-d3, also known as Vitamin B2-d3 or E101-d3, is the deuterium labeled form of Riboflavin. Riboflavin is an easily absorbed micronutrient that plays a key role in maintaining health in humans and other animals. This product is intended for research use only.
  • Tracer.
  • Internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes like deuterium into drug molecules for quantitation during drug development.
  • Potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07798