Learn More
Medchemexpress LLC Riboflavin phosphate sodium | 130-40-5 | 93.4% | 500 MG
SDP
Catalog No. 5000408276
Encompass_Preferred
Riboflavin phosphate sodium (FMN-Na) is a derivative of Riboflavin (vitamin B2), which is an essential nutrient for animals. It is utilized in research for progressive keratoconus, corneal ectasia, and irregular astigmatism. Riboflavine phosphate sodium is also described as a highly effective NAD+-recycling agent.
  • Derivative of Riboflavin (vitamin B2)
  • Essential nutrient for animals
  • Utilized in research for progressive keratoconus, corneal ectasia, and irregular astigmatism
  • Highly effective NAD+-recycling agent
Catalog No. 50-004-08276 Supplier Medchemexpress LLC Supplier No. HYB0964500MG
Riboflavin phosphate sodium (FMN-Na) is a derivative of Riboflavin (vitamin B2), which is an essential nutrient for animals. It is utilized in research for progressive keratoconus, corneal ectasia, and irregular astigmatism. Riboflavine phosphate sodium is also described as a highly effective NAD+-recycling agent.
  • Derivative of Riboflavin (vitamin B2)
  • Essential nutrient for animals
  • Utilized in research for progressive keratoconus, corneal ectasia, and irregular astigmatism
  • Highly effective NAD+-recycling agent