Learn More
Selleck Chemical LLC Riboflavin phosphate sodium S5107-1g
SDP
Catalog No. 503112170
Encompass_Preferred
Flavin mononucleotide (riboflavin-5 -phosphate FMN) is a biomolecule produced from riboflavin (vitamin B2) by the enzyme riboflavin kinase and functions as prosthetic group of various oxidoreductases including NADH dehydrogenase as well as cofactor in biological blue-light photo receptors
Catalog No. 50-311-2170 Supplier Selleck Chemical LLC Supplier No. S51071G
Flavin mononucleotide (riboflavin-5 -phosphate FMN) is a biomolecule produced from riboflavin (vitamin B2) by the enzyme riboflavin kinase and functions as prosthetic group of various oxidoreductases including NADH dehydrogenase as well as cofactor in biological blue-light photo receptors