Learn More
Medchemexpress LLC Riboflavin phosphate sodium hydrate | 6184-17-4 | 86.4% | 514.36 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013713100G
Encompass_Preferred
Riboflavin phosphate sodium hydrate is a derivative of Riboflavin (vitamin B2), an essential nutrient for animals. It is a very effective NAD+-recycling agent and can be used for research on progressive keratoconus, corneal ectasia, and irregular astigmatism.
  • Derivative of Riboflavin (vitamin B2)
  • Essential nutrient for animals
  • Used for research on progressive keratoconus, corneal ectasia, and irregular astigmatism
  • Very effective NAD+-recycling agent
Catalog No. 50-004-54739