Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Riboflavin USP Grade, MP Biomedicals™

Catalog No. MP210281325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210281325 25 g
MP210281305 5 g
MP210281380 100 g
MP210281390 500 g
MP210281391 1 kg
MP210281394 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP210281325 Supplier MP Biomedicals, Inc Supplier No. 0210281325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

USP Grade Orange Powder

  • A Vitamin/enzyme cofactor found in milk, eggs, malted barley, liver, kidney, heart, leafy vegetables
  • Used as a photoinitiator for polymerization of polyacrylamide gels
  • Richest natural source is yeast.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
Synonym Vitamin B2, Lactoflavin, Vitaming, 7,8-Dimethyl-10- (D-ribo-2,3,4,5- tetrahydroxypentyl) isoalloxazine
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point 290°C (decomposition)(literature)
Color Orange
Loss on Drying ≤1.5%
Quantity 25 g
Beilstein 97825
Solubility Information Soluble in 0.1 M Sodium Hydroxide (50mg/mL - clear, yellowish-orange solution); only slightly soluble in water.
Optical Rotation (c = 0.5, 0.05 M NaOH) -115° to -135°
Grade USP
Physical Form Powder
Assay >98%
RTECSNumber VJ1400000
storageNote1 Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.