Learn More
Medchemexpress LLC Riboflavin (Vitamin B2) | 83-88-5 | 99.9% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB045650G
Encompass_Preferred
Riboflavin, also known as Vitamin B2, is an orally active and easily absorbed micronutrient that serves as a precursor for flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). These act as coenzymes for various enzymatic reactions and are crucial for mediating electron transfer in biological oxidation-reduction processes. It is intended for research use only.
  • Precursor of FMN and FAD
  • Serves as coenzymes for numerous enzymatic reactions
  • Mediates electron transfer in biological oxidation-reduction reactions
  • Orally active and easily absorbed micronutrient
Catalog No. 50-004-05451