Learn More
LGC GENOMICS LLC (Lucigen) Riboguard RNase Inhibitor, 10,000 units @ 40 U/LRNA is precious. Protect it with this high quality, effective RNase inhibitor. Manufactured under ISO-13485.
Catalog No. NC0148451
Encompass
RiboGuard™ RNase Inhibitor is a recombinant protein that is the best defense against common RNases, including RNase A, RNase B, and RNase C.RiboGuard RNase Inhibitor is the best defence against common RNases, including RNase A, RNase B, and RNase C. This recombinant RNase inhibitor protein provides reliable protection of valuable RNA samples by binding strongly to RNases in a 1:1 ratio. Advanced purification protocols and extensive quality control tests ensure RiboGuard RNase Inhibitor is free of unwanted contaminants that can plague other commercially available preparations of RNase inhibitors.Applications: cDNA synthesis, RT-PCR, in vitro transcription and translationA potent affinity for RNases (Ki >10-14 M) ensures rapid inhibition even when trace amounts of RNase are present.Free of detectable RNase or DNase activity, and mammalian DNA.Does not interfere with enzymes commonly used to prepare or analyse RNA.Less sensitive to oxidation than traditional RNase inhibitors.
This item is not returnable. View return policy
Catalog No. NC0148451 Supplier LGC GENOMICS LLC Supplier No. RG90910K
RiboGuard™ RNase Inhibitor is a recombinant protein that is the best defense against common RNases, including RNase A, RNase B, and RNase C.RiboGuard RNase Inhibitor is the best defence against common RNases, including RNase A, RNase B, and RNase C. This recombinant RNase inhibitor protein provides reliable protection of valuable RNA samples by binding strongly to RNases in a 1:1 ratio. Advanced purification protocols and extensive quality control tests ensure RiboGuard RNase Inhibitor is free of unwanted contaminants that can plague other commercially available preparations of RNase inhibitors.Applications: cDNA synthesis, RT-PCR, in vitro transcription and translationA potent affinity for RNases (Ki >10-14 M) ensures rapid inhibition even when trace amounts of RNase are present.Free of detectable RNase or DNase activity, and mammalian DNA.Does not interfere with enzymes commonly used to prepare or analyse RNA.Less sensitive to oxidation than traditional RNase inhibitors.