Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ambion™ RiboMinus™ Eukaryote Kit for RNA-Seq

Catalog No. A1083708
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Preps (10 μg⁄prep)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1083708 8 Preps (10 μg⁄prep)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1083708 Supplier Ambion™ Supplier No. A1083708
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The RiboMinus™ Eukaryote Kit for RNA-Seq provides truly complete transcriptome isolation for next generation sequencing analysis.

The RiboMinus™ Eukaryote Kit for RNA-Seq provides truly complete transcriptome isolation for next generation sequencing analysis by offering a method for selective depletion of up to 99% of the 5S, 5.8S, 18S and 28S ribosomal RNA from total RNA versus poly-A or cap binding methods, which can introduce bias and select for only part of the transcriptome.

The RiboMinus™ Eukaryote Kit for RNA-Seq Offers:
• A simple, yet selective depletion procedure that uses a sequence-specific probe to capture rRNA
• Rapid and efficient removal of rRNA-probe complex using magnetic bead technology

Three Steps to an rRNA-free Preparation Suitable for Transcriptome Analysis
The removal of large ribosomal RNA using the RiboMinus™ kit is a reliable and robust three-step procedure. Hybridize the RNA sample to the RiboMinus™ Probe in a water bath or heat block, then remove probe-rRNA complexes using the RiboMinus™ magnetic beads included in the kit. Concentrate the depleted RNA preparation by ethanol precipitation, or by using the RiboMinus™ Concentration Module (Cat. No. K155005).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Each kit contains:
• 6 mL RiboMinus™ Magnetic Beads
• 80 μL RiboMinus™ Eukaryote Probe (15 pmol/μL)
• 10 mL Hybridization Buffer
• 12 mL DEPC-treated (RNase-free) water

Store at 4°C.

Isolation Technology Magnetic Bead
Sample Type Total RNA
Final Product Type Transcriptome RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Application) Microarray Analysis, Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Sequencing, Northern Blotting, cDNA Library Construction
No. of Reactions 8 Preps
Product Line RiboMinus
Quantity 8 Preps (10 μg⁄prep)
Sequencing Type Transcriptome Sequencing
Shipping Condition Wet Ice
Starting Material Amount 2-10 μg Total RNA
Target Transcriptome RNA
System Type RiboMinus
Yield 1 μg
Show More Show Less
What species have you tested with your RiboMinus Eukaryote Kit for RNA-Seq?

With regard to species specificity, the following organisms have 0 mismatches for the LSU/SSU probes (LSU (28S) / SSU (18S) Probe perfect matches). This means the probes will remove the 18S and 28S sequences from the following species.

- Species from the Land: Human, mouse, rat, frog, rabbit, cow, pig, chicken, Drosophila,* Caenorhabditis elegans, midge, mosquito, yeast (Saccharomyces cerevisiae, Schizosaccharomyces pombe)
- Species from the Sea: Sea squirt, coelacanth, lancelet, eel, salmon, sturgeon, ratfish, lamprey, brown plankton, zooplankton
- Plants: Arabidopsis thaliana, Brassica napus, rice, tomato, Humulus lupulus, Zea mays, wheat, soybean, pine, aspen

* The 28S and 18S probes in this kit respectively align 100% to the Drosophila 18S and 28S rRNA sequence. However, there are reports suggesting Drosophila (insect) 28S rRNA split into 2 smaller fragments co-migrating with 18S. The exact split site is not identified yet. But based on customer feedback, this kit does not remove the split 28S rRNA completely. We have not tested Drosophila samples with this kit.

The 5S and 5.8S sequences are more divergent, and thus the best probes possible have reduced species breadth:
- 5S: zero to 1 mismatch: human, mouse, rat, frog, chicken, Drosophila, C. elegans, A. thaliana, Z. mays
- 5.8S: zero mismatches: human, mouse, rat

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.