Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RiboMinus™ Eukaryote Kit v2

Catalog No. A15020
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15020 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15020 Supplier Invitrogen™ Supplier No. A15020
  • $1,448.00
    $1,389.65 / Each of 1

    Save $58.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The RiboMinus™ Eukaryote Kit v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA.

Features of the kit include:
• Seamless integration with RiboMinus™Concentration Module (available separately) to purify RNA after ribosomal RNA depletion
• 1.5-hour workflow
• Input range of 1-5 μg

The RiboMinus™ Eukaryote Kit v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA. When used with our library construction kits for sequencing, the RiboMinus™ Eukaryote Kit v2 provides maximum mapping to reference database with minimum bias, maintaining relative gene expression levels of even smaller RNA species. The resulting total RNA after depletion contains minimum contaminating rRNA with reduced variability.

The RiboMinus Eukaryote Kit v2 is designed for the removal of human, mouse, and rat rRNA. The RiboMinus™ Concentration Module (available separately) is used for RNA purification after rRNA depletion. For magnetic bead-based purification, please see the RiboMinus™ Eukaryote System v2. The RiboMinus Eukaryote Kit v2 has an RNA input range of 1-5 μg. For lower inputs (down to 100 ng), the Low Input RiboMinus™ Eukaryote System v2 is recommended.

The RiboMinus™ Eukaryote Kit v2 contains sufficient reagents to perform 12 ribosomal RNA depletions.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• RiboMinus™ Eukaryote Probe Mix, 2-20°C
• RiboMinus™ Magnetic Beads, 4°C
• 2 x Hybridization Buffer, room temperature

Isolation Technology Magnetic Bead
Species Human, Mouse, Rat
Final Product Type Transcriptome RNA
For Use With (Equipment) Ion PGM™ System, Ion Proton™ System, Illumina™ Genome Analyzer II (GA II)
Product Line RiboMinus
Quantity 12 reactions
Sequencing Type Transcriptome Sequencing
Starting Material Amount 1 - 5 μg total RNA
Workflow Step Sample Enrichment

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.