Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RiboMinus™ Plant Kit for RNA-Seq

Catalog No. A1083808
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Preps (10 μg⁄prep)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1083808 8 Preps (10 μg⁄prep)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1083808 Supplier Invitrogen™ Supplier No. A1083808
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

RiboMinus™ Plant Kit for RNA-Seq is the complete solution for transcriptome isolation and enrichment of the true whole transcriptome through selective depletion of ribosomal RNA in plant species.

RiboMinus™ Plant Kit for RNA-Seq is the complete solution for transcriptome isolation and enrichment of the true whole transcriptome through selective depletion of ribosomal RNA in plant species. This kit offers additional plastid depletion probes designed to target chloroplast rRNA not offered in the RiboMinus™ Eukaryote Kit for RNA-Seq critical for enhanced depletion of ribosomal RNA in plants. The RiboMinus™ Plant Kit for RNA-Seq contains all the components necessary to deplete ribosomal RNA from plant species. This improvement allows for up to 99.8% depletion of the major rRNA peaks in many plant species. The RiboMinus™ Plant Kit for RNA-Seq is recommended for wheat, maize, rice, barley, soybean, sugar cane, sorghum, potato, cassava, Arabidopsis, tomato, tobacco, poplar, legume, and moss. Additional plant species can be bioinformatically assessed by contacting Invitrogen.

Working with something other than human, mouse, or plant? Contact us with your species and sequence. Simply include your FASTA sequence in *.txt file format for the ribosomal sequences you would like to verify, with the subject line titled 'RiboMinus for (your organism here)', and we'll work with you to find the right probe for your RiboMinus™ rRNA depletion.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage

• Ribominus™ Plant Probe, 88 μL
• Hybridization Buffer, 11 mL
• Rnase Free Water, 13.2 mL
• RiboMinus™ Capture Beads, 6 mL
• User Manual

Isolation Technology Magnetic Bead
Sample Type Total RNA (Plant)
Final Product Type Transcriptome RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Sequencing, Northern Blotting, cDNA Library Construction
No. of Reactions 8 Preps
Product Line RiboMinus
Quantity 8 Preps (10 μg⁄prep)
Sequencing Type Transcriptome Sequencing
Shipping Condition Wet Ice
Starting Material Amount 2-10 μg Total RNA
Target Transcriptome RNA
System Type RiboMinus
Yield 1 μg
Show More Show Less
What type of plant samples can be used with the RiboMinus Plant Kit for RNA-Seq?

Plants that can be used with the RiboMinus Plant kit for RNA-Seq include: wheat, maize, rice, barley, soybean, sugar cane, sorghum, potato, cassava, Arabidopsis, tomato, tobacoo, poplar, legume, and moss.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.