Learn More
Apexbio Technology LLC Ribonic acid gamma lactone 5336-08-3 500mg
SDP
Catalog No. 5000088138
Encompass_Preferred
Ribonic acid gamma lactone (CAS 5336-08-3) is a small-molecule inhibitor targeting -galactosidase derived from Escherichia coli It is designed to inhibit enzyme activity thereby interfering with substrate hydrolysis in carbohydrate metabolic pathways Ribonic acid gamma lactone exerts its biological activity primarily through competitive binding to the active site of -galactosidase In in vitro studies ribonic acid gamma lactone demonstrates inhibitory activity with an inhibitory constant (Ki) of approximately 26 mM Based on these pharmacological properties ribonic acid gamma lactone holds research potential in studies examining enzyme-substrate interactions and carbohydrate metabolism mediated by bacterial -galactosidase
Catalog No. 50-000-88138 Supplier Apexbio Technology LLC Supplier No. B7913500
Ribonic acid gamma lactone (CAS 5336-08-3) is a small-molecule inhibitor targeting -galactosidase derived from Escherichia coli It is designed to inhibit enzyme activity thereby interfering with substrate hydrolysis in carbohydrate metabolic pathways Ribonic acid gamma lactone exerts its biological activity primarily through competitive binding to the active site of -galactosidase In in vitro studies ribonic acid gamma lactone demonstrates inhibitory activity with an inhibitory constant (Ki) of approximately 26 mM Based on these pharmacological properties ribonic acid gamma lactone holds research potential in studies examining enzyme-substrate interactions and carbohydrate metabolism mediated by bacterial -galactosidase