Learn More
New England Biolabs, Inc. Ribonucleotide Solution Set – 50 µmol of each
SDP
Catalog No. 504412
Encompass
The Ribonucleotide Solution Set (NTP Set) consists of four separate solutions of ATP, GTP, CTP and UTP, pH 7.5 as sodium salts. The NTPs are also available as a Ribonucleotide Solution Mix .
Catalog No. 50-441-2 Supplier New England Biolabs, Inc. Supplier No. N0450L
The Ribonucleotide Solution Set (NTP Set) consists of four separate solutions of ATP, GTP, CTP and UTP, pH 7.5 as sodium salts. The NTPs are also available as a Ribonucleotide Solution Mix .