Learn More
OriGene Ribonuclease 3 (RNASE3) CytoSection 25 Slides
SDP
Catalog No. NC3243656
Encompass_Preferred
Transient overexpression of RNASE3 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3243656 Supplier OriGene Supplier No. TS420569P5
Transient overexpression of RNASE3 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack