Learn More
Research Products International Corp Ribonuclease A, Bovine Pancreas, 1 Gram
SDP
Catalog No. 501368554
Encompass_Preferred
Molecular Biology Grade. Suitable for use in removing RNA in nucleic acid applications and where other enzymes are used or intact proteins need to be recovered. Chromatographically purified and supplied as a lyophilized powder.
Activity: ≥2,500 units/mg (dry weight)
Catalog No. 50-136-8554 Supplier Research Products International Corp Supplier No. R217501.0
Molecular Biology Grade. Suitable for use in removing RNA in nucleic acid applications and where other enzymes are used or intact proteins need to be recovered. Chromatographically purified and supplied as a lyophilized powder.
Activity: ≥2,500 units/mg (dry weight)