Learn More
Research Products International Corp Ribonuclease A, 100 MG
SDP
Catalog No. 501978096
Encompass_Preferred
Molecular Biology Grade. Suitable for use in removing RNA in nucleic acid applications and where other enzymes are used or intact proteins need to be recovered. Chromatographically purified and supplied as a lyophilized powder.
Activity: ≥2,500 units/mg (dry weight)
Catalog No. 50-197-8096 Supplier Research Products International Corp Supplier No. R217500.1
Molecular Biology Grade. Suitable for use in removing RNA in nucleic acid applications and where other enzymes are used or intact proteins need to be recovered. Chromatographically purified and supplied as a lyophilized powder.
Activity: ≥2,500 units/mg (dry weight)