Learn More
Worthington Biochemical Corporation Ribonuclease A, 200mg
SDP
Catalog No. NC1694192
Encompass
A chromatographically purified, lyophilized powder. Store at 2-8°C. PROTECT FROM MOISTURE. ≥2,500 units per mg dry weight
Catalog No. NC1694192 Supplier Worthington Biochemical Corporation Supplier No. LS003431
A chromatographically purified, lyophilized powder. Store at 2-8°C. PROTECT FROM MOISTURE. ≥2,500 units per mg dry weight