Learn More
Bioworld RNase A (Ribonuclease A), 500 mg
Catalog No. 50198992
Encompass_Preferred
RNase A Source: Bovine pancreas Activity (Kunitz, Protein @37°)- >60U/mgProtease freeUnit definition: one Kunitz unit of RNase activity causes the decrease in absorbance from A0 to Af per minute at 25°C at 400nm (yeast RNA substrate).Applications: For use in DNA analysis or used to depolymerize viscous cell extract.
Catalog No. 50-198-992 Supplier Bioworld Supplier No. 215600262
RNase A Source: Bovine pancreas Activity (Kunitz, Protein @37°)- >60U/mgProtease freeUnit definition: one Kunitz unit of RNase activity causes the decrease in absorbance from A0 to Af per minute at 25°C at 400nm (yeast RNA substrate).Applications: For use in DNA analysis or used to depolymerize viscous cell extract.