Learn More
Research Products International Corp Ribonuclease A 500 MG
SDP
Supplier:  Research Products International Corp R217500.5
Encompass_Preferred
Molecular Biology Grade. Suitable for use in removing RNA in nucleic acid applications and where other enzymes are used or intact proteins need to be recovered. Chromatographically purified and supplied as a lyophilized powder.
Activity: ≥2,500 units/mg (dry weight)
Catalog No. 50-197-7405