Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Ribonuclease A (Bovine)

Catalog No. 89960018 Shop All Abnova Corporation Products
Change view
Click to view available options
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-960-018 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-960-018 Supplier Abnova™ Supplier No. P5249
Only null left
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number P61823
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282340
Molecular Weight (g/mol) 13.7kDa
Name Ribonuclease A (Bovine)
Preparation Method Native protein chromatographically purified from Bovine Pancreas
Quantity 200 mg
Storage Requirements Store at 4°C on dry atmosphere.
Regulatory Status RUO
Gene Alias RNS1
Common Name RNASE1
Gene Symbol RNASE1
Assays Ribonuclease Assay; RNA removal during DNA isolation; RNA sequence analysis; RNase protection assays; RNA quantification or mapping.
Biological Activity ≥2,500 units per mg dry weight. One Kunitz unit is the amount of enzyme required to cause an increase in absorbance of 1.0 at 260 nm at 37°C (pH 5.0) when yeast rRNA is hydrolyzed to acid-soluble oligonucleotides. 50 units are equivalent to 1 Kunitz unit.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Bovine Pancreas
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.