Learn More
Alkali Scientific Ribonuclease A, Bovine Pancreas, 500 Mg
SDP
Catalog No. 5000661741
Encompass_Preferred
Ribonuclease A, derived from bovine pancreas, is a protein enzyme used for RNA digestion. This enzyme specifically cleaves RNA by hydrolyzing the phosphodiester bonds between nucleotides, making it an essential tool for RNA analysis and experimentation. The 500mg package is commonly used in RNA research to remove contaminating RNA during DNA purification or in ribonuclease assays. Ribonuclease A plays a pivotal role in molecular biology, particularly in the study of gene expression and the synthesis of RNA. It is also a valuable reagent in laboratory applications requiring selective RNA degradation, such as RNA isolation and purification processes in nucleic acid research.
Catalog No. 50-006-61741 Supplier Alkali Scientific Supplier No. RIB002CP
Ribonuclease A, derived from bovine pancreas, is a protein enzyme used for RNA digestion. This enzyme specifically cleaves RNA by hydrolyzing the phosphodiester bonds between nucleotides, making it an essential tool for RNA analysis and experimentation. The 500mg package is commonly used in RNA research to remove contaminating RNA during DNA purification or in ribonuclease assays. Ribonuclease A plays a pivotal role in molecular biology, particularly in the study of gene expression and the synthesis of RNA. It is also a valuable reagent in laboratory applications requiring selective RNA degradation, such as RNA isolation and purification processes in nucleic acid research.