Learn More
Sigma Aldrich Fine Chemicals Biosciences Ribonuclease A bovine250MG
Catalog No. 502902756
Encompass
Ribonucleases do not hydrolyze DNA because the DNA lacks 2-OH groups essential for the formation of cyclic intermediates. RNase can hydrolyze RNA from protein samples. Pancreatic RNase A specifically cleaves at the 3-side of pyrimidine (uracil or cytosine) phosphate bonds.
Catalog No. 50-290-2756 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R5000250MG
Ribonucleases do not hydrolyze DNA because the DNA lacks 2-OH groups essential for the formation of cyclic intermediates. RNase can hydrolyze RNA from protein samples. Pancreatic RNase A specifically cleaves at the 3-side of pyrimidine (uracil or cytosine) phosphate bonds.