Learn More
Crescent Chemical Co Inc RIBONUCLEASE A FRM BOVINE PANC
Supplier: Crescent Chemical Co Inc SE34388.01
Ribonuclease A from bovine pancreas min. 80 Kunitz units/mg lyophil. An endonuclease which specifically attacks pyrimidine sites (Py/pN) at the 3'-phosphate group. This preparation is salt-free, protease-free and chromatographically homogeneous.RNase A content: min. 90 % by ion exchange chromatography. DNa