Learn More
Crescent Chemical Co Inc RIBONUCLEASE A FRM BOVINE PANC
SDP
Supplier:  Crescent Chemical Co Inc SE34388.02
Encompass_Preferred
Ribonuclease A from bovine pancreas min. 80 Kunitz units/mg lyophil. An endonuclease which specifically attacks pyrimidine sites (Py/pN) at the 3'-phosphate group. This preparation is salt-free, protease-free and chromatographically homogeneous.RNase A content: min. 90 % by ion exchange chromatography. DNa
Catalog No. 50-248-101