Learn More
Sigma Aldrich Fine Chemicals Biosciences Ribonuclease A from bovine pancreas Type I-A, powder, >=60% RNase A basis (SDS-PAGE), >=50 Kunitz units/mg protein | 9001-99-4 | MFCD00132181 | 500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences R4875500MG
Encompass
Ribonuclease A from bovine pancreas Type I-A, powder, >=60% RNase A basis (SDS-PAGE), >=50 Kunitz units/mg protein | Purity: >=60% RNase A basis (SDS-PAGE) | 9001-99-4 | MFCD00132181 | 500MG
Catalog No. 50-177-9324