Learn More
Sigma Aldrich Fine Chemicals Biosciences Ribonuclease H RNase H fro
Catalog No. 502905752
Encompass
Ribonuclease H (RNase H) is a nonspecific endoribonuclease localized to the nucleus and cytoplasm. It is ubiquitously found and widely present among many organisms including viruses and human.
Catalog No. 50-290-5752 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10786357001
Ribonuclease H (RNase H) is a nonspecific endoribonuclease localized to the nucleus and cytoplasm. It is ubiquitously found and widely present among many organisms including viruses and human.