Learn More
Aldevron Ribonuclease Inhibitor, 2500ug
Catalog No. 503073863
Encompass_Preferred
2500ug dispense of Ribonuclease Inhibitor (Rnase Inhibitor). Ribonuclease Inhibitor is a potent, noncompetitive inhibitor of pancreatic-type ribonucleases including ribonucleases A, B and C.
Catalog No. 50-307-3863 Supplier Aldevron Supplier No. 91342500MCG
2500ug dispense of Ribonuclease Inhibitor (Rnase Inhibitor). Ribonuclease Inhibitor is a potent, noncompetitive inhibitor of pancreatic-type ribonucleases including ribonucleases A, B and C.