Learn More
Aldevron Ribonuclease Inhibitor, 660ug
Catalog No. 503073882
Encompass_Preferred
660ug dispense of Ribonuclease Inhibitor (Rnase Inhibitor) Ribonuclease Inhibitor is a potent, noncompetitive inhibitor of pancreatic-type ribonucleases including ribonucleases A, B and C.
Catalog No. 50-307-3882 Supplier Aldevron Supplier No. 9134660MCG
660ug dispense of Ribonuclease Inhibitor (Rnase Inhibitor) Ribonuclease Inhibitor is a potent, noncompetitive inhibitor of pancreatic-type ribonucleases including ribonucleases A, B and C.