Learn More
Medchemexpress LLC Ribonuclease T1 (animal free) | 9026-12-4 | 100 KU
SDP
Catalog No. 5000425649
Encompass_Preferred
Ribonuclease T1 (animal free) is an endonuclease that specifically degrades single-stranded RNA. It cleaves the phosphodiester bond between the 3′-guanosine residue and the 5′-OH group of the adjacent nucleoside, producing a 3′-GMP-terminated oligonucleotide, through a nucleoside 2′, 3′-cyclic phosphate intermediate. This product is animal-origin free and is for research use only.
  • Specifically degrades single-stranded RNA
  • Cleaves phosphodiester bond between 3′-guanosine residue and 5′-OH group
  • Produces 3′-GMP-terminated oligonucleotide
  • Animal-origin free
  • Suitable for research use
Catalog No. 50-004-25649 Supplier Medchemexpress LLC Supplier No. HYP0229A100KU
Ribonuclease T1 (animal free) is an endonuclease that specifically degrades single-stranded RNA. It cleaves the phosphodiester bond between the 3′-guanosine residue and the 5′-OH group of the adjacent nucleoside, producing a 3′-GMP-terminated oligonucleotide, through a nucleoside 2′, 3′-cyclic phosphate intermediate. This product is animal-origin free and is for research use only.
  • Specifically degrades single-stranded RNA
  • Cleaves phosphodiester bond between 3′-guanosine residue and 5′-OH group
  • Produces 3′-GMP-terminated oligonucleotide
  • Animal-origin free
  • Suitable for research use