Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribonucleic acid, MP Biomedicals™

Catalog No. ICN15202625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15202625 25 g
ICN15202680 100 g
ICN15202690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15202625 Supplier MP Biomedicals, Inc Supplier No. 0215202625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63231-63-0
MDL Number MFCD00132196
Synonym RNA
IUPAC Name Ribonucleic acid

Specifications

Color White
Quantity 25 g
Physical Form Powder
Chemical Name or Material Ribonucleic acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.