Learn More
Medchemexpress LLC Ribonucleic acid (yeast) | 63231-63-0 | MFCD00132196 | 92.4% | 250 MG
SDP
Catalog No. 5000282104
Encompass_Preferred
Ribonucleic acid (yeast) is a research-grade preparation of yeast-derived RNA supplied as a dry biochemical reagent for use in molecular biology and life-science research. It is commonly used as a substrate in enzymatic assays, as a control or blocking nucleic acid in molecular workflows, and as a general-purpose biological material for assay development and validation.
  • Provided as a dry material suitable for storage and handling.
  • Suitable as substrate for ribonuclease activity and enzymatic studies.
  • Useful as a control or blocking nucleic acid in molecular assays.
  • Available in multiple pack sizes to support both small-scale and bulk use.
  • Characterized with reported purity to aid reproducibility.
Catalog No. 50-002-82104 Supplier Medchemexpress LLC Supplier No. HYW250124250MG
Ribonucleic acid (yeast) is a research-grade preparation of yeast-derived RNA supplied as a dry biochemical reagent for use in molecular biology and life-science research. It is commonly used as a substrate in enzymatic assays, as a control or blocking nucleic acid in molecular workflows, and as a general-purpose biological material for assay development and validation.
  • Provided as a dry material suitable for storage and handling.
  • Suitable as substrate for ribonuclease activity and enzymatic studies.
  • Useful as a control or blocking nucleic acid in molecular assays.
  • Available in multiple pack sizes to support both small-scale and bulk use.
  • Characterized with reported purity to aid reproducibility.