Learn More
Sigma Aldrich Fine Chemicals Biosciences Ribonucleoside Triphosphat
Catalog No. 502905124
Encompass
Ribonucleoside Triphosphate Set consists of four separate vials of ATP CTP GTP and UTP. Each vial contains a 100 mM clear colorless SOLUTIONon of the lithium salt (pH 7).
Catalog No. 50-290-5124 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11277057001
Ribonucleoside Triphosphate Set consists of four separate vials of ATP CTP GTP and UTP. Each vial contains a 100 mM clear colorless SOLUTIONon of the lithium salt (pH 7).