Learn More
Sigma Aldrich Fine Chemicals Biosciences Ribonucleoside vanadyl com
Catalog No. 502901439
Encompass
Ribonucleoside vanadyl complexes are stable complexes formed between ribonucleotides. They are low molecular weight inhibitors of cellular ribonucleases (RNases) that are used for the isolation of RNA from cells.
Catalog No. 50-290-1439 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R33805ML
Ribonucleoside vanadyl complexes are stable complexes formed between ribonucleotides. They are low molecular weight inhibitors of cellular ribonucleases (RNases) that are used for the isolation of RNA from cells.