Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RiboPure™ RNA Purification Kit, yeast

Catalog No. am1926
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1926 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1926 Supplier Invitrogen™ Supplier No. AM1926
Only null left
Add to Cart
Add to Cart

For the isolation of extremely high-quality RNA from yeast without the use of enzymatic pretreatments that may alter gene expression profiles.

For the isolation of extremely high-quality RNA from yeast without the use of enzymatic pretreatments that may alter gene expression profiles. Each Invitrogen kit includes sufficient reagents for 50 purifications each from 107 to 108 yeast cells.
It has traditionally been difficult to isolate high-quality RNA from yeast because of problems disrupting the cells. Many protocols use enzymatic pretreatments that may affect gene expression patterns. The RiboPure-Yeast RNA Isolation Kit uses no enzymatic pretreatments and thus does not alter expression profiles. It combines a simple and efficient glass bead/one step phenol–mediated disruption using Invitrogen's Vortex Bead Beater Adapter followed by a glass fiber filter– based RNA purification step. After DNase treatment with the included DNA-free™ Reagents, the resulting RNA is exceptionally pure containing little to no genomic DNA (less than 0.05%). The kit has been used extensively at Invitrogen to isolate high-quality RNA from Saccharomyces cerevisiae, Schizosaccharomyces pombe, Pichia pastoris, and Candida albicans. The kit provides the highest yield of any method tested from each of these species.

  • Highest yields of total RNA available
  • No enzymatic pretreatment required
  • Includes DNA-free™ Reagents

Not high-throughput compatible (Manual)

Applications:
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Yeast

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 50 1.5-mL Screw Cap Tubes; store at room temperature
• 50 Filter Cartridges; room temperature
• 100 Collection Tubes (2 mL); room temperature
• 24 mL Lysis Buffer; 4°C
• 2.4 mL 10% SDS; 4°C
• 24 mL Phenol:Chloroform:IAA; 4°C
• 95 mL Binding Buffer; 4°C
• 35 mL Wash Solution 1; 4°C
• 50 mL Wash Solution 2/3 (concentrated); 4°C
• 5 mL Elution Solution; 4°C
• 80 g Zirconia Beads; -20°C
• 550 μL 10X DNase 1 Buffer; -20°C
• 200 μL DNase 1; -20°C
• 550 μL DNase Inactivation Reagent; -20°C
Isolation Technology Spin Column (Glass Fiber Filter)
Purification Time 1.5 hr. (Including DNase treatment)
Elution Volume 50–100 μL
Sample Type Yeast
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Shipping Condition Box 1: Dry Ice
Box 2: Room Temperature
Starting Material Amount Up to 108 cells
Yield Up to 300 μg
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.