Learn More
AdipoGen anti-Ribosomal Protein S3 (human), mAb (RP159-1) (100 µg)
SDP
Supplier:  AdipoGen AG20A0048C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human ribosomal protein S3. Detects a band of ~32kDa by Western blot. Isotype: Mouse IgG1kappa. Clone: RP159-1. Applications: ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. RpS3 is a component of the 40S ribosomal subunit and is an an essential but previously unkown subunit of NF-kappaB involved in the regulation of key genes in rapid cellular activation responses. RpS3 interacts with nm23-H1. The expression of rpS3 reduces the secretion of MMP-9 and the invasive metastatic potential in HT1080 cells. The phosphorylated ERK is reduced by the expression of rpS3.
Catalog No. 50-205-2848