Learn More
Cayman Chemical Ribosomal S6 Kinase 2 Polyclo
SDP
Catalog No. 502886883
Encompass_Preferred
Immunogen: Peptide from the C-terminal region of rat RSK2 • Host: rabbit • Species Reactivity: (+) rat RSK2 • Application(s): WB • RSKs 1-4 are downstream members of the ERK/MAPK cascade. Recent work suggests that RSK2 exerts a tonic regulation on G protein-coupled signaling.
Catalog No. 50-288-6883 Supplier Cayman Chemical Supplier No. 100094111 EA
Immunogen: Peptide from the C-terminal region of rat RSK2 • Host: rabbit • Species Reactivity: (+) rat RSK2 • Application(s): WB • RSKs 1-4 are downstream members of the ERK/MAPK cascade. Recent work suggests that RSK2 exerts a tonic regulation on G protein-coupled signaling.